Order Entry
Export
Orders LinkContactUsLinkComponent
Fenvalerate

Fenvalerate

Glass bottle for solids, 10 mg
Catalog # ACCUP-194N
Supplier:   AccuStandard
Fenvalerate
Catalog # ACCUP-194N
Supplier:  AccuStandard
CAS Number:  51630-58-1
Fenvalerate

Glass bottle for solids, 10 mg

CAS Number:  51630-58-1
Price
Inquire For Price
Log in for availability
1 * 10 mg
Product Details & Documents
  • CAS Number:51630-58-1
  • Formula:C₂₅H₂₂ClNO₃
  • MDL Number:MFCD00055324
  • EINECS:257-326-3
  • UN:2811
  • ADR:6.1,III
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    10 mg

Specifications