Ethylene glycol dimethacrylate (ethylene dimethacrylate) stabilised, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:97-90-5
:
Formula:C₁₀H₁₄O₄
MDL Number:MFCD00008590
:198.21 g/mol
:M-Clarity™ quality level = MQ200
:Stabilised with hydroquinone monomethyl ether (4-methoxyphenol, MEHQ)
EINECS:202-617-2