Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
1,4-Dioxane-2,5-dione
1,4-Dioxane-2,5-dione, Sigma-Aldrich®
1,4-Dioxane-2,5-dione, Sigma-Aldrich®
SIALG1796-25G
:
Merck
:
1,4-Dioxane-2,5-dione, Sigma-Aldrich®
SIALG1796-25G
:
Merck
:
Pack type:
Glass bottle for solids
Pk:
25 g
Seq:
3308
:
:
116.07 g/mol
:
M-Clarity™ quality level = MQ200
:
207-954-9
:
502-97-6
:
C₄H₄O₄
:
MFCD00081108