

Potassium tetrachloroaurate hydrate ≥99%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:27988-75-6
Formula:AuCl₄K·xH₂O
MDL Number:MFCD00043088
:377.88 g/mol (anhy)
Merck Index:14,7681
Melting Pt:357 °C (dec.)
Storage Temperature:Ambient
EINECS:237-190-1
UN:3260
ADR:8,III
:GHS05
:GHS07