Diuron (N-(3,4-dichlorophenyl)-N',N'-dimethylurea), Sigma-Aldrich®
Catalog # SIALD2425-100G
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:100 g
- Seq:3401
Specifications
About this item
:
:233.10 g/mol
EINECS:206-354-4
CAS Number:330-54-1
Formula:C₉H₁₀Cl₂N₂O
MDL Number:MFCD00018136