(S)-(+)-Methyl-3-hydroxybutyrate, Sigma-Aldrich®
Catalog # SIAL243167-1G
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:M-Clarity™ quality level = MQ100
:118.13 g/mol
EINECS:258-628-8
CAS Number:53562-86-0
Formula:C₅H₁₀O₃
MDL Number:MFCD00064461