Ethynyltributyltin 95%
Glass bottle, 2,5 g
Supplier: Thermo Fisher Scientific
Ethynyltributyltin 95%
Catalog # ACRO389180025
Supplier: Thermo Fisher Scientific
CAS Number: 994-89-8
Jump to:
Product Details & Documents
Synonyms: Ethynyltri-n-butylstannane, Tributylethynylstannane
- CAS Number:994-89-8
- Formula:C₁₄H₂₈Sn
- MDL Number:MFCD00009420
- Molecular Weight:315.09 g/mol
- Boiling Pt:70 °C (.2 mmHg)
- Flash Pt:73 °C
- Storage Temperature:Ambient
- UN:2788
- ADR:6.1,III
Documents
ACRO38918
SDS
Specifications
Specification Test Results
- Appearance:Clear colorless Liquid
- Infrared spectrum:Conforms
- Refractive index:1.4750 to 1.4770 (20°C, 589 nm)
- GC:≥94.0 %
Specifications
- Pack type:Glass bottle
- Pk:2,5 g



