Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
N-(2,4-Difluorophenyl)-2-chloroacetamide
N-(2,4-Difluorophenyl)-2-chloroacetamide
N-(2,4-Difluorophenyl)-2-chloroacetamide
N-(2,4-Difluorophenyl)-2-chloroacetamide
Supplier:
Thermo Scientific
Certificates
Expand image
N-(2,4-Difluorophenyl)-2-chloroacetamide
Supplier:
Thermo Scientific
About this item
CAS Number:
96980-65-3
:
Formula:
C₈H₆ClF₂NO
MDL Number:
MFCD00077473
:
205.5892 g/mol