N-(2,3-Dichlorophenyl)-2-chloroacetamide
Catalog # MAYBRF00037.50
Supplier: Thermo Scientific
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:50 g
- Seq:3368
Specifications
About this item
:
CAS Number:33560-47-3
Formula:C₈H₆Cl₃NO