

2-(4-Nitrophenyl)ethanol ≥98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:100-27-6
:
Formula:C₈H₉NO₃
MDL Number:MFCD00010202
:167.16 g/mol
Boiling Pt:177 °C (16 mmHg)
Melting Pt:59…62 °C
EINECS:202-835-8
:GHS06
:GHS07
Storage Temperature:Ambient