Order Entry
Export
ContactUsLinkComponent
4,4''-Dibromo-p-terphenyl, Sigma-Aldrich®
4,4''-Dibromo-p-terphenyl, Sigma-Aldrich®
Catalog # SIAL705411-5G
Supplier:  Merck
CAS Number:  
4,4''-Dibromo-p-terphenyl, Sigma-Aldrich®
Catalog # SIAL705411-5G
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    5 g
  • Seq:
    3173

Specifications

About this item

 :
 :388.1000 g/mol
CAS Number:17788-94-2
Formula:C₁₈H₁₂Br₂
MDL Number:MFCD00272889