Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Tris(2-methylphenyl)phosphine
Tris(2-methylphenyl)phosphine
Tris(2-methylphenyl)phosphine
Tris(2-methylphenyl)phosphine
Supplier:
Thermo Scientific
Certificates
Expand image
Tris(2-methylphenyl)phosphine
Supplier:
Thermo Scientific
About this item
CAS Number:
6163-58-2
:
Formula:
C₂₁H₂₁P
MDL Number:
MFCD00008514
:
304.37 g/mol
Boiling Pt:
187…192 °C (0,05 mmHg)
Melting Pt:
125,5 °C
EINECS:
228-193-9
:
GHS07