Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Ovens
Tube Furnaces
Accessories for rotary tube furnaces
Accessories for rotary tube furnaces
Accessories for rotary tube furnaces
Catalog # NABE691404552
Supplier:
NABERTHERM
Expand image
Accessories for rotary tube furnaces
Catalog # NABE691404552
Supplier:
NABERTHERM
Specifications
Description:
Quartz glass tube with pimple, 106×100×1540 mm, for Tmax. 1100 °C and 1300 °C models
Pk:
1
For:
RSR 120 - 500
Specifications
About this item
Product Details
Product Details