Tubing, rubber, porous
Cellular rubber, black.
- Flame resistant
- Covered with special skin
- Use for isolation
Synthetic natural rubber. Highly flexible and closed insulation material with low thermal conductivity.
Cellular rubber, black.
Synthetic natural rubber. Highly flexible and closed insulation material with low thermal conductivity.