2-Methyl-1,3-dithiane, Sigma-Aldrich®
Catalog # SIAL359130-5ML
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:5 mL
- Seq:2210
Specifications
About this item
:
:134.27 g/mol
EINECS:227-859-6
CAS Number:6007-26-7
Formula:C₅H₁₀S₂
MDL Number:MFCD00014652