2-Methyl-1,3-dithiane ≥98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:6007-26-7
:
Formula:C₅H₁₀S₂
MDL Number:MFCD00014652
:134.27 g/mol
Boiling Pt:78…80 °C (9 mmHg)
Density:1.212
Flash Pt:82 °C (179 °F)
Storage Temperature:Ambient
EINECS:227-859-6