Pesticide reference standard, Chlorfenvinphos (2-Chloro-1-(2,4 dichlorophenyl) vinyl diethyl phosphate, mixture of cis and trans isomers) 10 µg/ml in acetonitrile, Dr Ehrenstorfer
EHERL11290000AL
:
- Pack type:Glass bottle
- Pk:10 mL
- CAS No.:470-90-6
- Description:Chlorfenvinphos (2-Chloro-1-(2,4 dichlorophenyl) vinyl diethyl phosphate, mixture of cis and trans isomers) 10 µg/ml in acetonitrile
- Formula:C₁₂H₁₄Cl₃O₄P
- Description:Chlorfenvinphos (2-Chloro-1-(2,4 dichlorophenyl) vinyl diethyl phosphate, mixture of cis and trans isomers)