Pesticide reference standard, Carbofuran 100 µg/ml in methanol, Dr Ehrenstorfer
Catalog # EHERXA11010000ME
Supplier: EHRENSTORFER
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:1 mL
- CAS No.:1563-66-2
- Description:Carbofuran 100 µg/ml in methanol
- Formula:C₁₂H₁₅NO₃
- Description:Carbofuran