Herbicide reference standard, Cyanazine 100 µg/ml in acetonitrile, Dr Ehrenstorfer
EHERXA11790000AL
:
- Pack type:Glass bottle
- Pk:1 mL
- CAS No.:21725-46-2
- Description:Cyanazine 100 µg/ml in acetonitrile
- Formula:C₉H₁₃ClN₆
- Description:Cyanazine