Order Entry
Export
ContactUsLinkComponent
Carbaryl(1-naphthyl methylcarbamate)
Carbaryl(1-naphthyl methylcarbamate)
Catalog # EHERC10980000
Supplier:  EHRENSTORFER
CAS Number:  
Carbaryl(1-naphthyl methylcarbamate)
Catalog # EHERC10980000
Supplier:  EHRENSTORFER
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    0,25 g
  • Seq:
    3029

Specifications

About this item

 :
ADR:6.1,III
 :201.22 g/mol
EINECS:200-555-0
UN:2757
CAS Number:63-25-2
Formula:C₁₂H₁₁NO₂