Herbicide reference standard, Chlorpropham (Isopropyl 3-chlorocarbanilate) 10 µg/ml in acetonitrile, Dr Ehrenstorfer
Catalog # EHERL11580000AL
Supplier: EHRENSTORFER
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:10 mL
- CAS No.:101-21-3
- Description:Chlorpropham (Isopropyl 3-chlorocarbanilate) 10 µg/ml in acetonitrile
- Formula:C₁₀H₁₂ClNO₂
- Description:Chlorpropham (Isopropyl 3-chlorocarbanilate)