

Triethylenetetramine 60%, Technical Grade
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:112-24-3
:
Formula:C₆H₁₈N₄
MDL Number:MFCD00008169
:146.24 g/mol
Merck Index:14,9663
Boiling Pt:∼267 °C
Melting Pt:∼–35 °C
Density:0.982
Flash Pt:122 °C (252 °F)
Storage Temperature:Ambient
EINECS:203-950-6
UN:2259
ADR:8,II
:GHS05
:GHS07