Order Entry
Export
ContactUsLinkComponent
Pesticide reference standard, Dieldrin 100 µg/ml in cyclohexane, Dr Ehrenstorfer
Pesticide reference standard, Dieldrin 100 µg/ml in cyclohexane, Dr Ehrenstorfer
Catalog # EHERXA12590000CY
Supplier:  EHRENSTORFER
CAS Number:  
Pesticide reference standard, Dieldrin 100 µg/ml in cyclohexane, Dr Ehrenstorfer
Catalog # EHERXA12590000CY
Supplier:  EHRENSTORFER
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    1 mL
  • CAS No.:
    60-57-1
  • Description:
    Dieldrin 100 µg/ml in cyclohexane
  • Formula:
    C₁₂H₈Cl₆O
  • Description:
    Dieldrin

Specifications

About this item