

1,4-Dithiane-2,5-diol 96%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:40018-26-6
:
Formula:C₄H₈O₂S₂
MDL Number:MFCD00006659
:152.24 g/mol
Melting Pt:130 °C (dec.)
Storage Temperature:Refrigerator
EINECS:254-751-6
UN:2811
ADR:6.1,III
:GHS05
:GHS06
Specification Test Results
- Appearance:White to yellow Powder
- Infrared spectrum:Conforms
- GC:≥95.0 %
- HPLC:≥97.5 %