trans,trans-Muconic acid (trans,trans-1,3-butadiene-1,4-dicarboxylic acid), Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:3588-17-8
:
Formula:C₆H₆O₄
MDL Number:MFCD00002702
:142.11 g/mol
:M-Clarity™ quality level = MQ100
EINECS:222-724-8