
trans,trans-2,4-Decadienal 95% tech.
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:25152-84-5
:
Formula:C₁₀H₁₆O
MDL Number:MFCD00007007
:152.24 g/mol
Boiling Pt:114…116 °C (10 mmHg)
Density:0.85 g/cm³
Flash Pt:85 °C
EINECS:246-668-9
:GHS07
Storage Temperature:Refrigerator
Specification Test Results
- Appearance:Clear yellow Liquid
- Infrared spectrum:Conforms
- Refractive index:1.5140 to 1.5180 (20°C, 589 nm)
- GC:≥94 % (sum of isomers)