7-Hydroxy-1-tetralone
Catalog # MOLE22022921-1G
Supplier: Molekula
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:162.19 g/mol
CAS Number:22009-38-7
Formula:C₁₀H₁₀O₂
MDL Number:MFCD01312225