5-Hydroxy-1-tetralone
Catalog # MOLEM72619434
Supplier: Molekula
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:5 g
- Seq:3173
Specifications
About this item
:
:162.19 g/mol
EINECS:248-958-0
CAS Number:28315-93-7
Formula:C₁₀H₁₀O₂
MDL Number:MFCD00001693