Pesticide reference standard, Hexachlorocyclopentadiene 10 µg/ml in isooctane, Dr Ehrenstorfer
Catalog # EHERL14171000IO
Supplier: EHRENSTORFER
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:10 mL
- CAS No.:77-47-4
- Description:Hexachlorocyclopentadiene 10 µg/ml in isooctane
- Formula:C₅Cl₆
- Description:Hexachlorocyclopentadiene