Herbicide reference standard, Atrazine 100 µg/ml in acetonitrile, Dr Ehrenstorfer
Catalog # EHERXA10330000AL
Supplier: EHRENSTORFER
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:1 mL
- CAS No.:1912-24-9
- Description:Atrazine 100 µg/ml in acetonitrile
- Formula:C₈H₁₄ClN₅
- Description:Atrazine