Pesticide reference standard, β-Endosulfan 100 µg/ml in isooctane, Dr Ehrenstorfer
Catalog # EHERXA13122000IO
Supplier: EHRENSTORFER
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:1 mL
- CAS No.:33213-65-9
- Description:β-Endosulfan 100 µg/ml in isooctane
- Formula:C₉H₆Cl₆O₃S
- Description:β-Endosulfan