Pesticide reference standard, Tecnazene 100 µg/ml in isooctane, Dr Ehrenstorfer
Catalog # EHERXA17200000IO
Supplier: EHRENSTORFER
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:1 mL
- CAS No.:117-18-0
- Description:Tecnazene 100 µg/ml in isooctane
- Formula:C₆HCl₄NO₂
- Description:Tecnazene