Pesticide reference standard, Hexachlorobenzene 100 µg/ml in isooctane, Dr Ehrenstorfer
Catalog # EHERXA14160000IO
Supplier: EHRENSTORFER
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:1 mL
- CAS No.:118-74-1
- Description:Hexachlorobenzene 100 µg/ml in isooctane
- Formula:C₆Cl₆
- Description:Hexachlorobenzene