Holmium standard solution, 1,000 mg/L Ho in dilute nitric acid standard for ICP-MS
Catalog # ACCUICPMS24N0.1X1
Supplier: AccuStandard
CAS Number:
Specifications
- Concentration:1000 mg/L
- Matrix:Nitric acid dil.
- Pack type:Glass bottle
- Pk:100 mL
- Element:Holmium
- Symbol:Ho