Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Boron nitride
Boron nitride, rods, Length: 300 mm, Ø 12.7 mm
Boron nitride, rods, Length: 300 mm, Ø 12.7 mm
Boron nitride, rods, Length: 300 mm, Ø 12.7 mm
45912.KS
:
Thermo Fisher Scientific
:
Boron nitride, rods, Length: 300 mm, Ø 12.7 mm
45912.KS
:
Thermo Fisher Scientific
:
Pk:
1
Seq:
0001
:
10043-11-5
:
:
BN
:
MFCD00011317
:
24.82 g/mol
:
13,01336
:
2527 °C
:
2.15 g/cm³ (20 °C)
:
Ambient
:
233-136-6