1,1'-Thiocarbonyldiimidazole, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:6160-65-2
:
Formula:C₇H₆N₄S
MDL Number:MFCD00005289
:178.22 g/mol
Melting Pt:105…106 °C
:M-Clarity™ quality level = MQ100
Storage Temperature:Refrigerator
EINECS:228-183-4