

4-Nitrophthalonitrile ≥97%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:31643-49-9
:
Formula:C₈H₃N₃O₂
MDL Number:MFCD00040301
:173.13 g/mol
Melting Pt:136…137 °C
EINECS:250-748-9
:GHS06
:GHS07
Storage Temperature:Ambient