Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
5-Methyl-1,3-benzodioxole
5-Methyl-1,3-benzodioxole, Sigma-Aldrich®
5-Methyl-1,3-benzodioxole, Sigma-Aldrich®
SIAL284548-25ML
:
Merck
:
5-Methyl-1,3-benzodioxole, Sigma-Aldrich®
SIAL284548-25ML
:
Merck
:
Pack type:
Glass bottle
Pk:
25 mL
Seq:
2315
:
:
136.15 g/mol
:
M-Clarity™ quality level = MQ100
:
230-453-1
:
7145-99-5
:
C₈H₈O₂
:
MFCD00005833