GL adapters for barrels, BOLA
Adaptors made of PTFE/PP, for transition from female barrel thread R2" or Tri-Sure 2" to a GL 45 thread.
- Universal chemical resistance
- Temperature resistance from −200 to 250 °C
- Autoclavable and can be used under vacuum