Culture vials with cap, WHEATON®
Supplier: DWK Life Sciences
Certificates
About this item
These culture vials feature deep skirted screw caps to allow safer, more dependable handling when working with infectious materials. They are manufactured from 33 low extractable borosilicate glass that conforms to USP Type I and ASTM E 438 Type I, Class A requirements.
- Pre-attached deep skirted solid-top black phenolic screw cap with 14B rubber liner
Packaging: Vials are packaged in corrugated trays with partitions.