Affinity chromatography sorbents, Blue Trisacryl® M
Catalog # 555-0074
Supplier: Cytiva (Formerly Pall Lab)
CAS Number:
Specifications
- Ligand:Cibacron blue 3GA
- Binding capacity:≥10 mg/ml
- Max. pressure:3 bar
- Exclusion limit:107 Da
- Cat. no.:555-0074