Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
2,2'-Bithiophene
2,2'-Bithiophene, Sigma-Aldrich®
2,2'-Bithiophene, Sigma-Aldrich®
2,2'-Bithiophene, Sigma-Aldrich®
SIAL241636-10G
:
Merck
:
2,2'-Bithiophene, Sigma-Aldrich®
SIAL241636-10G
:
Merck
:
Pack type:
Glass bottle for solids
Pk:
10 g
Seq:
3236
:
:
166.27 g/mol
:
207-767-2
:
492-97-7
:
C₈H₆S₂
:
MFCD00005414