Order Entry
Export
Orders LinkContactUsLinkComponent
2,2'-Bithiophene

2,2'-Bithiophene

Glass bottle for solids, 5 g
Catalog # COBBOS-7500-5G
Supplier:   COMBI-BLOCKS
2,2'-Bithiophene
Catalog # COBBOS-7500-5G
Supplier:  COMBI-BLOCKS
CAS Number:  492-97-7
2,2'-Bithiophene

Glass bottle for solids, 5 g

CAS Number:  492-97-7
Price
Inquire For Price
Log in for availability
1 * 5 g
Product Details & Documents
Synonyms:  2,2′-Dithienyl, 2,2′-Bithienyl
  • CAS Number:492-97-7
  • Formula:C₈H₆S₂
  • MDL Number:MFCD00005414
  • Molecular Weight:166.27 g/mol
  • EINECS:207-767-2
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    5 g

Specifications