Order Entry
Export
Orders LinkContactUsLinkComponent
2,2'-Bithiophene ≥98%

2,2'-Bithiophene ≥98%

2,2'-Bithiophene ≥98%
Certificates
2,2'-Bithiophene ≥98%
2,2'-Bithiophene ≥98%

3 Options Available Below

Product Details & Documents

Purity: 98% CAS No.: 492-97-7 Molecular Formula: C8H6S2 Formula Weight: 166.27 Flash Point: 110°(230°F) Melting Point: 30-33° Boiling Point: 260° Beilstein Reference: 3039 MDL No.: MFCD00005414

Synonyms:  2,2′-Dithienyl, 2,2′-Bithienyl
  • CAS Number:492-97-7
  • Formula:C₈H₆S₂
  • MDL Number:MFCD00005414
  • Molecular Weight:166.27 g/mol
  • Boiling Pt:260 °C
  • Melting Pt:30…33 °C
  • Flash Pt:>110 °C (230 °F)
  • Storage Temperature:Ambient
  • EINECS:207-767-2
Product Family Options

Product Information