2,2-Dimethyl-1,3-dioxolane, Sigma-Aldrich®
Catalog # SIALD155624-25G
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:25 g
- Seq:3308
Specifications
About this item
:
:102.13 g/mol
:M-Clarity™ quality level = MQ200
EINECS:220-842-4
CAS Number:2916-31-6
Formula:C₅H₁₀O₂
MDL Number:MFCD00014106