Quinazoline
Glass bottle for solids, 5 g
Catalog # MOLEM38469977
Supplier: Molekula
Jump to:
Product Details & Documents
- CAS Number:253-82-7
- Formula:C₈H₆N₂
- MDL Number:MFCD00006712
- Molecular Weight:130.15 g/mol
- EINECS:205-965-3
Specifications
Specifications
- Pack type:Glass bottle for solids
- Pk:5 g



