Order Entry
Export
Orders LinkContactUsLinkComponent
4-Quinazolone

4-Quinazolone

Supplier:   Thermo Scientific
4-Quinazolone
Supplier:  Thermo Scientific
Certificates

3 Options Available Below

Product Details & Documents
Synonyms:  4-Hydroxyquinazoline, Quinazolin-4(3H)-one
  • CAS Number:491-36-1
  • Formula:C₈H₆N₂O
  • MDL Number:MFCD00511302
  • Molecular Weight:146.15 g/mol
  • EINECS:207-735-8
Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Glass bottle for solids
    1 g
    Log in for availability
    Log in for availability
    1 * 1 g
    1 * 1 g

  • Glass bottle for solids
    10 g
    Log in for availability
    Log in for availability
    1 * 10 g
    1 * 10 g

  • Glass bottle for solids
    50 g
    Log in for availability
    Log in for availability
    1 * 50 g
    1 * 50 g