1,3-Diethynylbenzene
Glass bottle for solids, 5 g
Catalog # MOLEM55367264
Supplier: Molekula
Jump to:
Product Details & Documents
- CAS Number:1785-61-1
- Formula:C₁₀H₆
- MDL Number:MFCD00041686
- Molecular Weight:126.16 g/mol
Specifications
Specifications
- Pack type:Glass bottle for solids
- Pk:5 g



