
1,4-Phenylenediacetic acid ≥97%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:7325-46-4
:
Formula:C₁₀H₁₀O₄
MDL Number:MFCD00004354
:194.19 g/mol
Melting Pt:252…256 °C
EINECS:230-801-2
:GHS07
Storage Temperature:Ambient