
1,4-Phenylenediacetic acid 97%
Catalog # ACRO130510050
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:5 g
- Seq:3173
Specifications
Specification Test Results
- Appearance:Light yellow to beige Powder
- Infrared spectrum:Conforms
- Melting point:252 to 257 °C
- HPLC:≥96.0 %
- Titration with NaOH:≥96.0 %
- pH:5 to 7.5 (1 % in water)
- Specific optical rotation:+75° to +105° (20 °C, 589 nm) (c=6, water)
- Specific viscosity:0.10 to 0.70 (1.0M in NaCl, 25 °C)
- Sulfur (S):17 to 20 %
Specification Test Results
About this item
:
:194.19 g/mol
Melting Pt:250…256 °C
EINECS:230-801-2
:GHS07
Storage Temperature:Ambient
CAS Number:7325-46-4
Formula:C₁₀H₁₀O₄
MDL Number:MFCD00004354